Learn More
Ace Glass, Inc. Flask, shake, 500mL, 4 bottom baffles, 38mm long bulbed neck, cs/12
SDP
Supplier:  Ace Glass, Inc. 389312
Encompass_Preferred
Flask, shake, 500mL, 4 bottom baffles, 38mm long bulbed neck, cs/12
Catalog No. 50-971-432