Learn More
Ace Glass, Inc. Flask, shake, 50mL, three deep baffles, 18mm OD long neck
SDP
Catalog No. 50971413
Encompass_Preferred
50mL cell culture shake flask with three outside edge baffles. The baffles are deeper that a standard flask to enhance mixing and oxygen transfer to media. The flask has an 18mm O.D. long neck to accomm ODate either plastic or stainless steel closures.
Catalog No. 50-971-413 Supplier Ace Glass, Inc. Supplier No. 388705
50mL cell culture shake flask with three outside edge baffles. The baffles are deeper that a standard flask to enhance mixing and oxygen transfer to media. The flask has an 18mm O.D. long neck to accomm ODate either plastic or stainless steel closures.