Learn More
Ace Glass, Inc. Flask, shake, 50mL, no baffles, 18mm OD long neck, cs/24
SDP
Catalog No. 50971406
Encompass_Preferred
50mL shake style culture flask without baffles. Flask has an 18mm O.D. long neck, and accomm ODates either plastic or stainless steel closures.
Catalog No. 50-971-406 Supplier Ace Glass, Inc. Supplier No. 388410
50mL shake style culture flask without baffles. Flask has an 18mm O.D. long neck, and accomm ODates either plastic or stainless steel closures.