Learn More
Ace Glass, Inc. Flask, shake, 50mL, three side baffles, 18mm OD long neck, cs/12
SDP
Catalog No. 50971400
Encompass_Preferred
50mL shake flask with triple side baffles for cell culture applications. Flask has an 18mm O.D. long style straight neck to accomm ODate plastic and stainless steel closures. The flask is triple baffled on the bottom edge to increase mixing and oxygen transfer to the media.
Catalog No. 50-971-400 Supplier Ace Glass, Inc. Supplier No. 388303
50mL shake flask with triple side baffles for cell culture applications. Flask has an 18mm O.D. long style straight neck to accomm ODate plastic and stainless steel closures. The flask is triple baffled on the bottom edge to increase mixing and oxygen transfer to the media.