Learn More
Chemglass Life Sciences Spinner Flask, 1L, Adjustable Hanging Bar, Complete
SDP
Catalog No. 50143796
Encompass_Preferred
Adjustable hanging bar spinner flasks are designed for growth of suspension cultures.The system includes a flat bottom flask with two angled sidearms and a stir bar with glass shaft that adjusts through the compression fitting in the cap.
Catalog No. 50-143-796 Supplier Chemglass Life Sciences Supplier No. CLS14451L
Adjustable hanging bar spinner flasks are designed for growth of suspension cultures.The system includes a flat bottom flask with two angled sidearms and a stir bar with glass shaft that adjusts through the compression fitting in the cap.