Learn More
Chemglass Life Sciences BioProcess Spinner Flask, 3, 000mL, Internal Impeller, Complete
SDP
Catalog No. 50143730
Encompass_Preferred
MicroCarrier spinner flasks promote optimal growth of suspension and microcarrier cultures.Indented bottom (100mL thru 1000mL) prevents accumulation of microcarrier beads under the impeller paddle.The 3L version does not have a dimple.The complete system includes a flask with two angled sidearms and a PTFE paddle assembly that attaches underneath the cap decreasing the risk of contamination.
Catalog No. 50-143-730 Supplier Chemglass Life Sciences Supplier No. CLS14503L
MicroCarrier spinner flasks promote optimal growth of suspension and microcarrier cultures.Indented bottom (100mL thru 1000mL) prevents accumulation of microcarrier beads under the impeller paddle.The 3L version does not have a dimple.The complete system includes a flask with two angled sidearms and a PTFE paddle assembly that attaches underneath the cap decreasing the risk of contamination.