Learn More
Chemglass Life Sciences BioProcess Spinner Flask, 250mL, Dimpled Bottom, Complete
SDP
Catalog No. 50143727
Encompass_Preferred
Microcarrier spinner flasks promote optimal growth of suspension and microcarrier cultures.Indented bottoms, 100mL thru 1000mL, prevent accumulation of microcarrier beads under the impeller paddle.Please Note: The 6L and 8L Versions Do Not Have Dimpled Bottoms.The complete system includes a flask with two angled sidearms and a PTFE paddle with glass shaft that adjusts trough the compression fitting in the cap.
Catalog No. 50-143-727 Supplier Chemglass Life Sciences Supplier No. CLS1430250
Microcarrier spinner flasks promote optimal growth of suspension and microcarrier cultures.Indented bottoms, 100mL thru 1000mL, prevent accumulation of microcarrier beads under the impeller paddle.Please Note: The 6L and 8L Versions Do Not Have Dimpled Bottoms.The complete system includes a flask with two angled sidearms and a PTFE paddle with glass shaft that adjusts trough the compression fitting in the cap.