Learn More
Chemglass Life Sciences BioProcess Spinner Flask, 500mL, Flat Bottom, Complete
SDP
Catalog No. 50143838
Encompass_Preferred
Flat bottom spinner flasks promote optimal growth of suspension cultures.The complete system includes a flask with two angled sidearms and a PTFE paddle with glass shaft that adjusts through the compression fitting in the cap.
Catalog No. 50-143-838 Supplier Chemglass Life Sciences Supplier No. CLS1430F500
Flat bottom spinner flasks promote optimal growth of suspension cultures.The complete system includes a flask with two angled sidearms and a PTFE paddle with glass shaft that adjusts through the compression fitting in the cap.