Learn More
Ace Glass, Inc. Flask, storage, 250mL, bottom hose connection, #6 stopper size beaded neck, cs/6
SDP
Catalog No. 50971441
Encompass_Preferred
250 mL media storage flask with 1/2" bottom hose connection. Ideal for use with aseptic filling bells for transferring medias to various other flasks and vessels. Beaded rim neck takes standard cotton plugs or stoppers.
Catalog No. 50-971-441 Supplier Ace Glass, Inc. Supplier No. 390603
250 mL media storage flask with 1/2" bottom hose connection. Ideal for use with aseptic filling bells for transferring medias to various other flasks and vessels. Beaded rim neck takes standard cotton plugs or stoppers.