Learn More
Ace Glass, Inc. Flask, storage, 2L, bottom hose connection, #9 stopper size beaded neck, cs/3
SDP
Catalog No. 50971443
Encompass_Preferred
2L media storage flask with 1/2" bottom hose connection. Ideal for use with aseptic filling bells for transferring medias to various other flasks and vessels. Beaded rim neck takes standard cotton plugs or stoppers.
Catalog No. 50-971-443 Supplier Ace Glass, Inc. Supplier No. 390609
2L media storage flask with 1/2" bottom hose connection. Ideal for use with aseptic filling bells for transferring medias to various other flasks and vessels. Beaded rim neck takes standard cotton plugs or stoppers.