Learn More
Ace Glass, Inc. Flask, storage, 4L, bottom hose connection, #10 stopper size beaded neck, cs/3
SDP
Catalog No. 50971444
Encompass_Preferred
4L media storage flask with 1/2" bottom hose connection. Ideal for use with aseptic filling bells for transferring medias to various other flasks and vessels. Beaded rim neck takes standard cotton plugs or stoppers.
Catalog No. 50-971-444 Supplier Ace Glass, Inc. Supplier No. 390611
4L media storage flask with 1/2" bottom hose connection. Ideal for use with aseptic filling bells for transferring medias to various other flasks and vessels. Beaded rim neck takes standard cotton plugs or stoppers.