Learn More
Ace Glass, Inc. Flask, storage, 500mL, bottom hose connection, #7 stopper size beaded neck, cs/6
SDP
Catalog No. 50978220
Encompass_Preferred
500 mL media storage flask with 1/2" bottom hose connection. Ideal for use with aseptic filling bells for transferring medias to various other flasks and vessels. Beaded rim neck takes standard cotton plugs or stoppers.
Catalog No. 50-978-220 Supplier Ace Glass, Inc. Supplier No. 390605
500 mL media storage flask with 1/2" bottom hose connection. Ideal for use with aseptic filling bells for transferring medias to various other flasks and vessels. Beaded rim neck takes standard cotton plugs or stoppers.