Learn More
Ace Glass, Inc. Flask, storage, 6L, bottom hose connection, #11 stopper size beaded neck, cs/2
SDP
Catalog No. 50978221
Encompass_Preferred
6L media storage flask with 1/2" bottom hose connection. Ideal for use with aseptic filling bells for transferring medias to various other flasks and vessels. Beaded rim neck takes standard cotton plugs or stoppers.
Catalog No. 50-978-221 Supplier Ace Glass, Inc. Supplier No. 390613
6L media storage flask with 1/2" bottom hose connection. Ideal for use with aseptic filling bells for transferring medias to various other flasks and vessels. Beaded rim neck takes standard cotton plugs or stoppers.