Learn More
Chemglass Life Sciences Flask, Straus, 500mL, Airfree, Schlenk, 24/40 Joint, 0-4mm 1-Arm Valve
SDP
Catalog No. 501310982
Encompass_Preferred
Flask, Straus, 500mL, Airfree, Schlenk, 24/40 Joint, 0-4mm 1-Arm Valve. Round bottom storage flask which can maintain dry, grease-free and deoxygenated solvent storage for long periods of time. Unique design permits filling of the flask using a 24/40 inlet off of a vacuum line and recovery of the solvent via a cannula or syringe through the valve body.
Catalog No. 50-131-0982 Supplier Chemglass Life Sciences Supplier No. AF052501
Flask, Straus, 500mL, Airfree, Schlenk, 24/40 Joint, 0-4mm 1-Arm Valve. Round bottom storage flask which can maintain dry, grease-free and deoxygenated solvent storage for long periods of time. Unique design permits filling of the flask using a 24/40 inlet off of a vacuum line and recovery of the solvent via a cannula or syringe through the valve body.