Learn More
Electron Microscopy Sciences Dumont Flask Tong Style ER13TSC, 12 mm ID Grasp, Inox 08, Matt Finish
SDP
Catalog No. 501931114
Encompass_Preferred
Tool adapted for chemistry, radiochemistry, and radiation metrology meeting the highest quality standards. Features a long handle and covered tips for antistatic protection.
I.D.: 12 mm
Material: Inox 08
Finish: Matt
OAL: 250
Tip thickness: 12 mm
Catalog No. 50-193-1114 Supplier Electron Microscopy Sciences Supplier No. 7672310
Tool adapted for chemistry, radiochemistry, and radiation metrology meeting the highest quality standards. Features a long handle and covered tips for antistatic protection.
I.D.: 12 mm
Material: Inox 08
Finish: Matt
OAL: 250
Tip thickness: 12 mm