Learn More
Bellco Glass, Inc. Flask Tray, 125mL
SDP
Catalog No. 501946148
Encompass_Preferred
Pre-mounted stainless steel clamps for bellco Erlenmeyer flask. Flask tray fits on top of the standard tray platform. Includes clamps.. MADE IN THE USA Flask positions: 25 Dimensions: 25x25cm Size: 125ml
Catalog No. 50-194-6148 Supplier Bellco Glass, Inc. Supplier No. 774412525
Pre-mounted stainless steel clamps for bellco Erlenmeyer flask. Flask tray fits on top of the standard tray platform. Includes clamps.. MADE IN THE USA Flask positions: 25 Dimensions: 25x25cm Size: 125ml