Learn More
Bellco Glass, Inc. Flask Tray, 1L
SDP
Catalog No. 501946145
Encompass_Preferred
Pre-mounted stainless steel clamps for bellco Erlenmeyer flask. Flask tray fits on top of the standard tray platform. Includes clamps.. MADE IN THE USA Flask positions: 9 Size:1l
Catalog No. 50-194-6145 Supplier Bellco Glass, Inc. Supplier No. 774410009
Pre-mounted stainless steel clamps for bellco Erlenmeyer flask. Flask tray fits on top of the standard tray platform. Includes clamps.. MADE IN THE USA Flask positions: 9 Size:1l