Learn More
Bellco Glass, Inc. Flask Tray,500mL
SDP
Catalog No. 501946162
Encompass_Preferred
Pre-mounted stainless steel clamps for bellco Erlenmeyer flask. Flask tray fits on top of the standard tray platform. Includes clamps. Flask positions: 4 Dimensions: 25x25cm Size: 500ml. MADE IN THE USA
Catalog No. 50-194-6162 Supplier Bellco Glass, Inc. Supplier No. 774450004
Pre-mounted stainless steel clamps for bellco Erlenmeyer flask. Flask tray fits on top of the standard tray platform. Includes clamps. Flask positions: 4 Dimensions: 25x25cm Size: 500ml. MADE IN THE USA