Learn More
Bellco Glass, Inc. Flask Tray 500 mL
SDP
Catalog No. 501946163
Encompass_Preferred
Pre-mounted stainless steel clamps for bellco Erlenmeyer flask. Flask tray fits on top of the standard tray platform. Includes clamps. Flask positions: 9 Dimensions: 51x51cm Size: 500ml. MADE IN THE USA
Catalog No. 50-194-6163 Supplier Bellco Glass, Inc. Supplier No. 774450009
Pre-mounted stainless steel clamps for bellco Erlenmeyer flask. Flask tray fits on top of the standard tray platform. Includes clamps. Flask positions: 9 Dimensions: 51x51cm Size: 500ml. MADE IN THE USA