Learn More
Ace Glass, Inc. Flask, trypsinizing, 1.5L, baffled, #8 stopper size beaded neck cs/2
SDP
Supplier:  Ace Glass, Inc. 389721
Encompass_Preferred
1.5L trypsinizing flask with beaded neck and long side baffles. Ideal for converting homogenous tissue samples into cell suspensions by digestion of connective tissue. Heavy beaded neck accepts either cotton plugs or stoppers. The extra-deep baffles enhance vigorous agitation.
Catalog No. 50-971-437