Learn More
Chemglass Life Sciences Flask, Trypsinizing, 2L, 158mm Dia x 290mm OAH
SDP
Supplier:  Chemglass Life Sciences CLS205802
Encompass_Preferred
Flask, Trypsinizing, 2L, 158mm Dia x 290mm OAH. Manufactured from Type 1 Class A borosilicate glass with FOUR deep side baffles for optimum agitation during trypsinization. Flask comes with beaded, break-resistant lips and 50mm OD necks. Sold 1 per case.
Catalog No. 50-131-1297