Learn More
Ace Glass, Inc. Flask, trypsinizing, 300mL, baffled, #6 stopper size beaded neck, cs/3
SDP
Catalog No. 50978218
Encompass_Preferred
300mL trypsinizing flask with beaded neck and long side baffles. Ideal for converting homogenous tissue samples into cell suspensions by digestion of connective tissue. Heavy beaded neck accepts either cotton plugs or stoppers. The extra-deep baffles enhance vigorous agitation.
Catalog No. 50-978-218 Supplier Ace Glass, Inc. Supplier No. 389714
300mL trypsinizing flask with beaded neck and long side baffles. Ideal for converting homogenous tissue samples into cell suspensions by digestion of connective tissue. Heavy beaded neck accepts either cotton plugs or stoppers. The extra-deep baffles enhance vigorous agitation.