Learn More
Ace Glass, Inc. Flask, trypsinizing, 3L, baffled, #10 stopper size beaded /2
SDP
Catalog No. 50971438
Encompass_Preferred
3L trypsinizing flask with beaded neck and long side baffles. Ideal for converting homogenous tissue samples into cell suspensions by digestion of connective tissue. Heavy beaded neck accepts either cotton plugs or stoppers. The extra-deep baffles enhance vigorous agitation.
Catalog No. 50-971-438 Supplier Ace Glass, Inc. Supplier No. 389725
3L trypsinizing flask with beaded neck and long side baffles. Ideal for converting homogenous tissue samples into cell suspensions by digestion of connective tissue. Heavy beaded neck accepts either cotton plugs or stoppers. The extra-deep baffles enhance vigorous agitation.