Learn More
Chemglass Life Sciences Flask, Trypsinizing, 4L, 205mm Dia x 360mm OAH
SDP
Catalog No. 501311298
Encompass_Preferred
Flask, Trypsinizing, 4L, 205mm Dia x 360mm OAH. Manufactured from Type 1 Class A borosilicate glass with FOUR deep side baffles for optimum agitation during trypsinization. Flask comes with beaded, break-resistant lips and 50mm OD necks. Sold 1 per case.
Catalog No. 50-131-1298 Supplier Chemglass Life Sciences Supplier No. CLS205804
Flask, Trypsinizing, 4L, 205mm Dia x 360mm OAH. Manufactured from Type 1 Class A borosilicate glass with FOUR deep side baffles for optimum agitation during trypsinization. Flask comes with beaded, break-resistant lips and 50mm OD necks. Sold 1 per case.