Learn More
Ace Glass, Inc. Flask, trypsinizing, 700mL, baffled, #7 stopper size beaded neck, cs/2
SDP
Catalog No. 50971436
Encompass_Preferred
700mL trypsinizing flask with beaded neck and long side baffles. Ideal for converting homogenous tissue samples into cell suspensions by digestion of connective tissue. Heavy beaded neck accepts either cotton plugs or stoppers. The extra-deep baffles enhance vigorous agitation.
Catalog No. 50-971-436 Supplier Ace Glass, Inc. Supplier No. 389719
700mL trypsinizing flask with beaded neck and long side baffles. Ideal for converting homogenous tissue samples into cell suspensions by digestion of connective tissue. Heavy beaded neck accepts either cotton plugs or stoppers. The extra-deep baffles enhance vigorous agitation.