Learn More
Ace Glass, Inc. Flask, trypsinizing, 75mL, baffled, with sidearm & neck screwcaps, cs/3
SDP
Catalog No. 50978219
Encompass_Preferred
75mL trypsinizing flask for converting of homogenous tissue samples into cell suspensions by digestion of connective tissue. Side port has dam below the side arm to prevent undigested tissue from being poured out while decanting cells. Flask has GPI thread top and side arm cap. The three extra deep, extra long side baffles greatly enhance vigorous agitation.
Catalog No. 50-978-219 Supplier Ace Glass, Inc. Supplier No. 390232
75mL trypsinizing flask for converting of homogenous tissue samples into cell suspensions by digestion of connective tissue. Side port has dam below the side arm to prevent undigested tissue from being poured out while decanting cells. Flask has GPI thread top and side arm cap. The three extra deep, extra long side baffles greatly enhance vigorous agitation.