Learn More
Hach Company Flask, Volumetric, Class A, Certified, 100 mL
Supplier:  Hach Company 2636642
Encompass
Individually serialized glass flask supplied with Certificate of Graduation Accuracy and traceable to NIST Standards. Class A tolerances meet requirements of ASTM E438 and ASTM E288. Graduation ring on neck of flask. Supplied with ground glass stopper. (2603213 size 13 replacement stoppers).
Catalog No. 50-199-6428