Learn More
Hach Company Flask, Volumetric, Class A, Certified, 500 mL
Supplier:  Hach Company 2636649
Encompass
Individually serialized glass flask supplied with Certificate of Graduation Accuracy and traceable to NIST Standards. Class A tolerances meet requirements of ASTM E438 and ASTM E288. Graduation ring on neck of flask. Supplied with ground glass stopper. (replacement stoppers size 19).
Catalog No. 50-199-6430