Learn More
Chemglass Life Sciences 1000mL Volumetric Flask, Class A, #22 Outer Stopper Neck, Glass Stopper
SDP
Catalog No. CG160009
Encompass_Preferred
1000mL Volumetric Flask, Class A, #22 Outer Stopper Neck, Glass Stopper. Flasks are manufactured to conform to Federal Specifications NNN-F-289 for Type 1, Class “A” unserialized flasks and ASTM E-288. Supplied complete with one solid glass pennyhead stopper. For additional stoppers, see CG-3018.
Catalog No. CG160009 Supplier Chemglass Life Sciences Supplier No. CG160009
1000mL Volumetric Flask, Class A, #22 Outer Stopper Neck, Glass Stopper. Flasks are manufactured to conform to Federal Specifications NNN-F-289 for Type 1, Class “A” unserialized flasks and ASTM E-288. Supplied complete with one solid glass pennyhead stopper. For additional stoppers, see CG-3018.