Learn More
Chemglass Life Sciences Flask, Volumetric, 250mL, PMP, Class A, 19/26, Tolerance: +-0.15mL
SDP
Catalog No. 501311076
Encompass_Preferred
Flask, Volumetric, 250mL, PMP, Class A, 19/26, Tolerance: ¿0.15mL. Polymethylpentene (PMP) volumetric flasks are manufactured to conform to Class ¿A¿ standards per DIN EN ISO 1042. The flasks are packed with a printed lot number and batch certificate. The polymethylpentene flask offers a unique combination of transparency, heat and chemical resistance. Flasks have a scored graduation line, with coloring added for visibility. Supplied complete with matching polypropylene standard taper joint stoppers. Flask can be autoclaved at 121¿C.
Catalog No. 50-131-1076 Supplier Chemglass Life Sciences Supplier No. CG161207
Flask, Volumetric, 250mL, PMP, Class A, 19/26, Tolerance: ¿0.15mL. Polymethylpentene (PMP) volumetric flasks are manufactured to conform to Class ¿A¿ standards per DIN EN ISO 1042. The flasks are packed with a printed lot number and batch certificate. The polymethylpentene flask offers a unique combination of transparency, heat and chemical resistance. Flasks have a scored graduation line, with coloring added for visibility. Supplied complete with matching polypropylene standard taper joint stoppers. Flask can be autoclaved at 121¿C.