Learn More
Chemglass Life Sciences Flask, Volumetric, 100mL, PP, Class B, GL-18, Tolerance: +-0.20mL
SDP
Catalog No. 501311082
Encompass_Preferred
Flask, Volumetric, 100mL, PP, Class B, GL-18, Tolerance: ¿0.20mL. Polypropylene(PP) volumetric flasks are manufactured to conform to Class ¿B¿ standards per DIN EN ISO 1042. The transparent polypropylene flasks are an excellent break-resistant alternative to glass. Flasks have a scored graduation line, with coloring added for visibility. Supplied complete with matching polypropylene screw cap.
Catalog No. 50-131-1082 Supplier Chemglass Life Sciences Supplier No. CG161305
Flask, Volumetric, 100mL, PP, Class B, GL-18, Tolerance: ¿0.20mL. Polypropylene(PP) volumetric flasks are manufactured to conform to Class ¿B¿ standards per DIN EN ISO 1042. The transparent polypropylene flasks are an excellent break-resistant alternative to glass. Flasks have a scored graduation line, with coloring added for visibility. Supplied complete with matching polypropylene screw cap.