Learn More
Chemglass Life Sciences Flask, Volumetric, 50mL, PP, Class B, GL-18, Tolerance: +-0.12mL
SDP
Catalog No. 501311081
Encompass_Preferred
Flask, Volumetric, 50mL, PP, Class B, GL-18, Tolerance: ¿0.12mL. Polypropylene(PP) volumetric flasks are manufactured to conform to Class ¿B¿ standards per DIN EN ISO 1042. The transparent polypropylene flasks are an excellent break-resistant alternative to glass. Flasks have a scored graduation line, with coloring added for visibility. Supplied complete with matching polypropylene screw cap.
Catalog No. 50-131-1081 Supplier Chemglass Life Sciences Supplier No. CG161304
Flask, Volumetric, 50mL, PP, Class B, GL-18, Tolerance: ¿0.12mL. Polypropylene(PP) volumetric flasks are manufactured to conform to Class ¿B¿ standards per DIN EN ISO 1042. The transparent polypropylene flasks are an excellent break-resistant alternative to glass. Flasks have a scored graduation line, with coloring added for visibility. Supplied complete with matching polypropylene screw cap.