Learn More
Hach Company Flask, Volumetric Class A, Glass, 100 mL
Supplier:  Hach Company 1457442
Encompass
Calibrated “To Contain” and manufactured to ASTM E438, “Glass in Laboratory Apparatus” requirements.
Catalog No. 50-199-5081