Learn More
Chemglass Life Sciences 100mL Volumetric Flask, Class A, #13 Outer Stopper Neck, PE Stopper
SDP
Catalog No. CG160205
Encompass_Preferred
100mL Volumetric Flask, Class A, #13 Outer Stopper Neck, PE Stopper. Flasks are manufactured to conform to Federal Specifications NNN-F-289 for Type 1, Class “A” unserialized flasks and ASTM E-288. Supplied complete with one closed bottom polyethylene stopper. For additional stoppers, see CG-8740.
Catalog No. CG160205 Supplier Chemglass Life Sciences Supplier No. CG160205
100mL Volumetric Flask, Class A, #13 Outer Stopper Neck, PE Stopper. Flasks are manufactured to conform to Federal Specifications NNN-F-289 for Type 1, Class “A” unserialized flasks and ASTM E-288. Supplied complete with one closed bottom polyethylene stopper. For additional stoppers, see CG-8740.