Learn More
Vee Gee Volumetric Flask, Amber, Class A, Glass Stopper, 1000 mL
Catalog No. NC2340782
Encompass_Preferred
The SIBATA Amber Class A Volumetric Flask is constructed of low-actinic amber glass, providing protection for materials that are sensitive to light. It features machine-blown body and heavy-beaded, heavy-tubing neck that is tooled for standard taper glass stoppers.• Constructed of borosilicate glass with a low coefficient of expansion• Designed """"To Contain"""" with a temperature of calibration at 20°C• Flask has a large marking square and includes a glass stopper• Meets ASTM E-288 Class A Specifications & USP Standards for Volumetric Flasks
Catalog No. NC2340782 Supplier Vee Gee Supplier No. 2307A1000
The SIBATA Amber Class A Volumetric Flask is constructed of low-actinic amber glass, providing protection for materials that are sensitive to light. It features machine-blown body and heavy-beaded, heavy-tubing neck that is tooled for standard taper glass stoppers.• Constructed of borosilicate glass with a low coefficient of expansion• Designed """"To Contain"""" with a temperature of calibration at 20°C• Flask has a large marking square and includes a glass stopper• Meets ASTM E-288 Class A Specifications & USP Standards for Volumetric Flasks