Learn More
Hach Company Flask, Volumetric, Polymethylpentene, 100 mL
Supplier:  Hach Company 2099542
Encompass
Durable polymethylpentene flask with polypropylene screw-top closure. Calibrated “to contain/deliver” at 20°C.
Catalog No. 50-199-5378