Learn More
Chemglass Life Sciences 100 +/- 0.10mL Capacity, #16 Stopper Size, Approx 180mm Height with Stopper
SDP
Catalog No. CG1616100
Encompass_Preferred
Volumetric Flask, 100 +/- 0.10mL Capacity, #16 Stopper Size, Approx 180mm Height with Stopper. Low actinic red stained glass provides protection to light sensitive solutions. Allowing less than 1% transmission below 400nm and 5% from 400-600nm. Wide mouth design facilitates mixing, pipette access, sampling and filling. Unique Flat Bottom Interior permits the use of a standard stir bar. Large permanent white letters make the capacity clearly visible. Flasks are calibrated "To Contain" at Class A tolerances, per ASTM E288 specifications. Supplied complete with a glass stopper.
Catalog No. CG1616100 Supplier Chemglass Life Sciences Supplier No. CG1616100
Volumetric Flask, 100 +/- 0.10mL Capacity, #16 Stopper Size, Approx 180mm Height with Stopper. Low actinic red stained glass provides protection to light sensitive solutions. Allowing less than 1% transmission below 400nm and 5% from 400-600nm. Wide mouth design facilitates mixing, pipette access, sampling and filling. Unique Flat Bottom Interior permits the use of a standard stir bar. Large permanent white letters make the capacity clearly visible. Flasks are calibrated "To Contain" at Class A tolerances, per ASTM E288 specifications. Supplied complete with a glass stopper.