Learn More
Chemglass Life Sciences Flask, 2800mL, Fernbach, 70mm Screw Thread, Without Baffles, Solid Cap, Approx OD x Height (mm): 210 x 230
SDP
Catalog No. 50143699
Encompass_Preferred
Flask, 2800mL, Fernbach, 70mm Screw Thread, Without Baffles, Solid Cap, Approx OD x Height (mm): 210 x 230. 2800mL flasks are designed for culturing organisms that require a large surface area to volume ratio.
Catalog No. 50-143-699 Supplier Chemglass Life Sciences Supplier No. CLS202170
Flask, 2800mL, Fernbach, 70mm Screw Thread, Without Baffles, Solid Cap, Approx OD x Height (mm): 210 x 230. 2800mL flasks are designed for culturing organisms that require a large surface area to volume ratio.