Learn More
Chemglass Life Sciences Flask, 2800mL, Fernbach, 70mm Screw Thread, With 3 Side Baffles, Vented Cap, Approx OD x Height (mm): 210 x 230
SDP
Catalog No. 50143815
Encompass_Preferred
Flask, 2800mL, Fernbach, 70mm Screw Thread, With 3 Side Baffles, Vented Cap, Approx OD x Height (mm): 210 x 230. 2800mL flask with 3 baffles on the lower outside edge of the flask to help oxygen transfer to the culture medium.
Catalog No. 50-143-815 Supplier Chemglass Life Sciences Supplier No. CLS202372
Flask, 2800mL, Fernbach, 70mm Screw Thread, With 3 Side Baffles, Vented Cap, Approx OD x Height (mm): 210 x 230. 2800mL flask with 3 baffles on the lower outside edge of the flask to help oxygen transfer to the culture medium.