Learn More
Chemglass Life Sciences Flask, Fernbach, 2800mL, GL-45 Thread, Side and Bottom Baffles, Vented Cap, Approx OD x Height (mm): 210 x 230
SDP
Catalog No. 50143739
Encompass_Preferred
Flask, Fernbach, 2800mL, GL-45 Thread, Side and Bottom Baffles, Vented Cap, Approx OD x Height (mm): 210 x 230. Flasks are designed for culturing organisms that require a large surface area to volume ratio. Flask has 3 baffles on the lower outside edge and 3 baffles on the bottom surface to help oxygen transfer to the culture medium.
Catalog No. 50-143-739 Supplier Chemglass Life Sciences Supplier No. CLS202633
Flask, Fernbach, 2800mL, GL-45 Thread, Side and Bottom Baffles, Vented Cap, Approx OD x Height (mm): 210 x 230. Flasks are designed for culturing organisms that require a large surface area to volume ratio. Flask has 3 baffles on the lower outside edge and 3 baffles on the bottom surface to help oxygen transfer to the culture medium.