Learn More
Cole-Parmer Pyrex 5100-500 Brand 5100 flask; 500 mL, pack of 6
Supplier:  Cole-Parmer 3455603
Encompass_Preferred
These wide mouth flasks are especially designed for use as a titration flask. For your convenience, these flasks are graduated to show the approximate capacity and have an extra large marking spot.
Catalog No. NC9942397