Learn More
Chemglass Life Sciences Flask, Shake, 2L, 6 Bottom Baffles, 70mm Vented Cap, Approx OD x Height (mm): 170 x 270
SDP
Catalog No. 50143810
Encompass_Preferred
Flask, Shake, 2L, 6 Bottom Baffles, 70mm Vented Cap, Approx OD x Height (mm): 170 x 270. Flask has 6 bottom baffles to maximize oxygen transfer for bacterial, yeast and other cultures that require large amounts of oxygen for optimal growth. The 70mm vented cap has a 0.20µm PVDF hydrophobic membrane to allow for sterile gas exchange while preventing contamination. A silicone liner ensures a tight seal between the cap and flask. The flasks, cap and liners are all fully autoclavable.
Catalog No. 50-143-810 Supplier Chemglass Life Sciences Supplier No. CLS205404
Flask, Shake, 2L, 6 Bottom Baffles, 70mm Vented Cap, Approx OD x Height (mm): 170 x 270. Flask has 6 bottom baffles to maximize oxygen transfer for bacterial, yeast and other cultures that require large amounts of oxygen for optimal growth. The 70mm vented cap has a 0.20µm PVDF hydrophobic membrane to allow for sterile gas exchange while preventing contamination. A silicone liner ensures a tight seal between the cap and flask. The flasks, cap and liners are all fully autoclavable.