Learn More
Braintree Scientific Flat Bottom, 3.25"" dia x 8"", 250-500gm rat
SDP
Catalog No. 501954813
Encompass
Economy Flat Bottomed Restrainers. Individual semi cylindrical plastic restrainers in small, medium and large. Each unit has an open slot on the bottom (for intra-peritoneal injections), a hole on the top (for the neck), and a slotted tail piece (for intravenous injections). Front slots provide ample ventilation. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4813 Supplier Braintree Scientific Supplier No. FBL
Economy Flat Bottomed Restrainers. Individual semi cylindrical plastic restrainers in small, medium and large. Each unit has an open slot on the bottom (for intra-peritoneal injections), a hole on the top (for the neck), and a slotted tail piece (for intravenous injections). Front slots provide ample ventilation. **Sold for research purposes only and not for human use**