Learn More
Foxx Life Sciences Borosil® Flat Bottom Boiling Flask with Short Neck and Interchangeable Joint (24/29), 1L (1000mL), 2/CS
Catalog No. 501720480
Encompass_Preferred
Borosil® Flat Bottom Boiling Flasks have uniform wall thickness for even heat distribution. Flasks are stress free giving them mechanical strength as well as thermal shock resistance. Borosil® Boiling Flasks are heat-resistant. Built to withstand the demands of the modern lab, each flask has a flame-polished, Interchangable Joint, and an enamel area for marking. Available from 50mL to 2L. Flasks comply with ISO 4797. Some options of 250mL,500mLand 1L options not covered in ISO. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices.
Catalog No. 50-172-0480 Supplier Foxx Life Sciences Supplier No. 4100C29
Borosil® Flat Bottom Boiling Flasks have uniform wall thickness for even heat distribution. Flasks are stress free giving them mechanical strength as well as thermal shock resistance. Borosil® Boiling Flasks are heat-resistant. Built to withstand the demands of the modern lab, each flask has a flame-polished, Interchangable Joint, and an enamel area for marking. Available from 50mL to 2L. Flasks comply with ISO 4797. Some options of 250mL,500mLand 1L options not covered in ISO. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices.