Learn More
Bellco Glass, Inc. Flat Bottom AHB Spinner , 1000mL No Ctr Cap
SDP
Catalog No. 501945147
Encompass_Preferred
Adjustable Hanging Bar Spinner Flask without Bottom Dimple. MADE IN THE USAPTFE impeller assembly rides on glass bearing for friction free operation. 25 & 50mL have black phenolic rubberlined center and sidearm caps.100mL-300mL have polypropylene caps, silicone gasket and pouring rings accumulation of microcarrier beads under impeller. 1000mL. Center cap: 100mm, side arm 45mm. Dimensions: 135 OD x 240 OAH
Catalog No. 50-194-5147 Supplier Bellco Glass, Inc. Supplier No. 196711000
Adjustable Hanging Bar Spinner Flask without Bottom Dimple. MADE IN THE USAPTFE impeller assembly rides on glass bearing for friction free operation. 25 & 50mL have black phenolic rubberlined center and sidearm caps.100mL-300mL have polypropylene caps, silicone gasket and pouring rings accumulation of microcarrier beads under impeller. 1000mL. Center cap: 100mm, side arm 45mm. Dimensions: 135 OD x 240 OAH