Learn More
Bellco Glass, Inc. Flat Bottom AHB Spinner ,25mL No Ctr Cap
SDP
Catalog No. 501945142
Encompass_Preferred
Adjustable spinner height. Ptfe impeller assembly rides on glass bearings for friction free operation. 25ml & 50ml have black phenolic rubber lined caps. 100ml-300ml have polypropylene caps, silicone gasket and pouring rings. MADE IN THE USA. Size: 25ml Dimensions: 38mm OX 120mm TALL. CENTER NECK: 38mm. SIDEARMS: 18mm. BLACK PHENOLIC RUBBER LINED SCAPS. GRADUATIONS: 5ml TO 25ml.
Catalog No. 50-194-5142 Supplier Bellco Glass, Inc. Supplier No. 196710025
Adjustable spinner height. Ptfe impeller assembly rides on glass bearings for friction free operation. 25ml & 50ml have black phenolic rubber lined caps. 100ml-300ml have polypropylene caps, silicone gasket and pouring rings. MADE IN THE USA. Size: 25ml Dimensions: 38mm OX 120mm TALL. CENTER NECK: 38mm. SIDEARMS: 18mm. BLACK PHENOLIC RUBBER LINED SCAPS. GRADUATIONS: 5ml TO 25ml.