Learn More
Bellco Glass, Inc. Flat Bottom AHB Spinner , 500mL No Ctr Cap
SDP
Catalog No. 501945146
Encompass_Preferred
Adjustable Hanging Bar Spinner Flask without Bottom Dimple. MADE IN THE USAPTFE impeller assembly rides on glass bearing for friction free operation. 25 & 50mL have black phenolic rubberlined center and sidearm caps.100mL-300mL have polypropylene caps, silicone gasket and pouring rings accumulation of microcarrier beads under impeller. 500mL. Center cap: 100mm, side arm 45mm. Dimensions: 110 OD x 185 OAH
Catalog No. 50-194-5146 Supplier Bellco Glass, Inc. Supplier No. 196710500
Adjustable Hanging Bar Spinner Flask without Bottom Dimple. MADE IN THE USAPTFE impeller assembly rides on glass bearing for friction free operation. 25 & 50mL have black phenolic rubberlined center and sidearm caps.100mL-300mL have polypropylene caps, silicone gasket and pouring rings accumulation of microcarrier beads under impeller. 500mL. Center cap: 100mm, side arm 45mm. Dimensions: 110 OD x 185 OAH