Learn More
Bellco Glass, Inc. Flat Bottom AHB Spinner Complete, 100mL
SDP
Catalog No. 501945137
Encompass_Preferred
•Designed for the efficient growth of suspension cells at low speeds. MADE IN THE USA•Flask Specifications:o100 mL flat bottom, Type 1 Class A BorosilicateoDimensions: 63.5mm OD x 135mm OAHoTwo 32mm Sidearms angled 55º offset centeroOne 70mm Screw thread center neckoGraduations: 20mL to 100mL, every 20mLoIncludes two 32mm blue PP screw caps and pouring rings•Impeller Assembly Specifications:oConsists of PTFE coated magnet and white Teflon magnet holder•Glass Shaft Specifications:oType 1 Class A BorosilicateoDimensions: 7mm OD x 155mm long•Cap Assembly Specifications:oConsists of one 70mm blue PP screw cap with 16.2 mm hole for black compression fitting, 70mm white silicone liner with 15.8mm hole, 7mm black compression fitting
Catalog No. 50-194-5137 Supplier Bellco Glass, Inc. Supplier No. 196700100A
•Designed for the efficient growth of suspension cells at low speeds. MADE IN THE USA•Flask Specifications:o100 mL flat bottom, Type 1 Class A BorosilicateoDimensions: 63.5mm OD x 135mm OAHoTwo 32mm Sidearms angled 55º offset centeroOne 70mm Screw thread center neckoGraduations: 20mL to 100mL, every 20mLoIncludes two 32mm blue PP screw caps and pouring rings•Impeller Assembly Specifications:oConsists of PTFE coated magnet and white Teflon magnet holder•Glass Shaft Specifications:oType 1 Class A BorosilicateoDimensions: 7mm OD x 155mm long•Cap Assembly Specifications:oConsists of one 70mm blue PP screw cap with 16.2 mm hole for black compression fitting, 70mm white silicone liner with 15.8mm hole, 7mm black compression fitting