Learn More
Bellco Glass, Inc. Adjustable spinner height. Ptfe impeller assembly rides on glass bearings for friction free operation. 25ml & 50ml have black phenolic rubber lined caps. 100ml-300ml have polypropylene caps, silicone gasket and pouring rings. Flask has flat bottom (no
SDP
Catalog No. 501945136
Encompass_Preferred
Adjustable spinner height. Ptfe impeller assembly rides on glass bearings for friction free operation. 25ml & 50ml have black phenolic rubber lined caps. 100ml-300ml have polypropylene caps, silicone gasket and pouring rings. Flask has flat bottom (no
Catalog No. 50-194-5136 Supplier Bellco Glass, Inc. Supplier No. 196700050
Adjustable spinner height. Ptfe impeller assembly rides on glass bearings for friction free operation. 25ml & 50ml have black phenolic rubber lined caps. 100ml-300ml have polypropylene caps, silicone gasket and pouring rings. Flask has flat bottom (no