Learn More
Grainger Wiremold OFR Series Flat Elbow, Steel, Black, 1 ea
Catalog No. 502758441
Encompass_Preferred
This flat elbow is designed for use within the OFR Series overfloor raceway system to provide connectivity transitions in commercial applications. It offers a low-profile, ADA compliant solution for routing power, communications, and A/V cables over existing floor surfaces.
  • Provides a secure 90-degree turn in the raceway path
  • Features a durable steel construction in black
  • Meets ADA accessibility guidelines for standard 4.8
  • Tamper-resistant and installer-friendly design
  • Paintable surface to match interior decor
  • cETLus listed for safety and performance compliance
Catalog No. 50-275-8441 Supplier Grainger Supplier No. 15A810
This flat elbow is designed for use within the OFR Series overfloor raceway system to provide connectivity transitions in commercial applications. It offers a low-profile, ADA compliant solution for routing power, communications, and A/V cables over existing floor surfaces.
  • Provides a secure 90-degree turn in the raceway path
  • Features a durable steel construction in black
  • Meets ADA accessibility guidelines for standard 4.8
  • Tamper-resistant and installer-friendly design
  • Paintable surface to match interior decor
  • cETLus listed for safety and performance compliance